2-(4-Pyridyl)benzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Pyridinyl)benzaldehyde, 2-(Pyridin-4-yl)benzaldehyde

A324697-100MG A324697-250MG A324697-1G A324697-5G
103825-376EA 169 USD
103825-376 103836-320 76724-028 76724-030 77306-318 77306-320
2-(4-Pyridyl)benzaldehyde 97%
2-(4-Pyridyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₉NO
MW: 183.2100 g/mol
Boiling Pt: 342.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD02684098
CAS Number: 176526-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR