3-(2-Hydroxyethyl)pyridine 95%

Supplier: Ambeed
Warning

A260399-1G A260399-250MG A260399-5G A260399-10G
103812-494EA 28 USD
103812-494 103852-376 77091-964 77193-810 77304-792 77746-427
3-(2-Hydroxyethyl)pyridine 95%
3-(2-Hydroxyethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₉NO
Boiling Pt: 242.2 °C (760 mm Hg)
Flash Pt: 100.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00831047
CAS Number: 6293-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR