2-(3-Pyridyl)benzaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Pyridin-3-yl)benzaldehyde

A237313-250MG A237313-1G
103811-146EA 33 USD
103811-146 103850-924 76722-350 77090-004
2-(3-Pyridyl)benzaldehyde 95%
2-(3-Pyridyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₉NO
Boiling Pt: 356.8 °C (760 mm Hg)
Melting Pt: 83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02684097
CAS Number: 176690-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR