2-(Pyridin-3-yl)acetic acid hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Pyridyl-3-acetic acid hydrochloride, 3-Pyridylacetic acid hydrochloride

A172627-25G A172627-100G A172627-10G A172627-500G A172627-5G
103845-024EA 6 USD
103845-024 77082-230 77191-462 77191-464 77191-466 77301-206
2-(Pyridin-3-yl)acetic acid hydrochloride 95%
2-(Pyridin-3-yl)acetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈ClNO₂
MW: 173.60 g/mol
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00012819
CAS Number: 6419-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR