2-(Pyrid-2-yloxy)benzaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Pyridin-2-yloxy)benzaldehyde

A497733-1G A497733-100MG A497733-5G
103828-270EA 84 USD
103828-270 103839-350 76843-318 76878-028 77307-604
2-(Pyrid-2-yloxy)benzaldehyde 95%
2-(Pyrid-2-yloxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₉NO₂
MW: 199.205 g/mol
Boiling Pt: 324.9 °C (760 mm Hg)
Flash Pt: 150.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09025852
CAS Number: 141580-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR