2-(Pyridin-2-yl)pyrimidin-4-amine 97%

Supplier: Ambeed
Warning

A246445-100MG A246445-250MG A246445-1G
103811-712EA 50 USD
103811-712 103851-546 77013-892 77013-894 77304-390
2-(Pyridin-2-yl)pyrimidin-4-amine 97%
2-(Pyridin-2-yl)pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₉H₈N₄
MW: 172.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08704306
CAS Number: 61310-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR