2-(2-Hydroxyprop-2-yl)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Pyridin-2-yl)propan-2-ol

A131849-1G A131849-250MG A131849-100MG
103801-646EA 91 USD
103801-646 77298-516 77549-570 77747-941
2-(2-Hydroxyprop-2-yl)pyridine 97%
2-(2-Hydroxyprop-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.179 g/mol
Boiling Pt: 226.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00209259
CAS Number: 37988-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR