2-(2-Pyridyl)-2-propylamine 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Pyridin-2-yl)propan-2-amine

A250150-250MG A250150-100MG
103811-914EA 104 USD
103811-914 103851-752 76976-670 77612-414
2-(2-Pyridyl)-2-propylamine 95%
2-(2-Pyridyl)-2-propylamine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂
MW: 136.194 g/mol
Boiling Pt: 212.7±15.0 °C (760 mm Hg)
Flash Pt: 103.1 °C
Storage Temperature: Ambient
MDL Number: MFCD08729302
CAS Number: 52568-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR