p-Tolylacetic acid 98%

Supplier: Ambeed
Warning

A349592-25G A349592-1000G A349592-100G A349592-10G A349592-500G
103837-014EA 5 USD
103837-014 76840-110 77251-516 77251-518 77251-520 77251-522
p-Tolylacetic acid 98%
p-Tolylacetic acid

Precaución: for Research Use Only

Formula: (C₆H₄(CH₃))CH₂COOH
Boiling Pt: 265…267 °C (760 mm Hg)
Melting Pt: 90…93 °C
Flash Pt: 173.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004353
CAS Number: 622-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR