2-(2-Propynyloxy)tetrahydro-2H-pyran 97%

Supplier: Ambeed
Danger

Synonyms: Tetrahydro-2-(2-propynyloxy)-2H-pyran, 2-Propargyloxane, 2-(Prop-2-yn-1-yloxy)tetrahydro-2H-pyran, Propargyl 2-tetrahydropyranyl ether, 2-(2-Propynyloxy)tetrahydropyran

A209604-10G A209604-1G A209604-5G A209604-100G
103817-948EA 6 USD
103817-948 103848-636 76770-448 77086-984 77086-986 77598-316
2-(2-Propynyloxy)tetrahydro-2H-pyran 97%
2-(2-Propynyloxy)tetrahydro-2H-pyran

Precaución: for Research Use Only

Formula: C₈H₁₂O₂
Boiling Pt: 63…65 °C (760 mm Hg)
Flash Pt: 59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006604
CAS Number: 6089-04-9
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR