2-(Piperidin-1-yl)benzaldehyde 97%

Supplier: Ambeed
Warning

A413908-1G A413908-25G A413908-5G A413908-10G
103827-264EA 14 USD
103827-264 103838-308 76841-864 76841-866 76841-868 77668-870
2-(Piperidin-1-yl)benzaldehyde 97%
2-(Piperidin-1-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO
MW: 189.2500 g/mol
Boiling Pt: 323.8 °C (760 mm Hg)
Flash Pt: 124.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03419311
CAS Number: 34595-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR