2-Piperazin-1-yl-4-(trifluoromethyl)pyrimidine 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Piperazin-1-yl)-4-(trifluoromethyl)pyrimidine

A283676-5G A283676-100MG A283676-250MG
103813-622EA 71 USD
103813-622 103853-572 76837-284 77277-014 77277-016
2-Piperazin-1-yl-4-(trifluoromethyl)pyrimidine 95%
2-Piperazin-1-yl-4-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₉H₁₁F₃N₄
MW: 232.21 g/mol
Boiling Pt: 331.3 °C (760 mm Hg)
Melting Pt: 50…52 °C
Flash Pt: 154.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00203912
CAS Number: 179756-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR