2-(2-Methylphenyl)ethanol 98%

Supplier: Ambeed
Warning

Synonyms: 2-(o-Tolyl)ethanol

A246652-1G A246652-25G A246652-5G A246652-10G
103811-722EA 10 USD
103811-722 103851-556 77090-850 77090-852 77090-854 77276-198
2-(2-Methylphenyl)ethanol 98%
2-(2-Methylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₂O
MW: 136.1900 g/mol
Boiling Pt: 243.5 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00044241
CAS Number: 19819-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR