2-(N-Methylamino)-3-hydroxymethylpyridine 98%

Supplier: Ambeed
Warning

A460859-250MG A460859-1G
103827-916EA 6 USD
103827-916 103838-994 76774-218 76842-872
2-(N-Methylamino)-3-hydroxymethylpyridine 98%
2-(N-Methylamino)-3-hydroxymethylpyridine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.1700 g/mol
Boiling Pt: 312.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06254696
CAS Number: 32399-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR