2-(1-Naphthalene)ethanol 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Naphthalen-1-yl)ethanol, 2-(1-Naphthyl)ethanol

A207559-100G A207559-10G A207559-500G A207559-1G A207559-25G A207559-5G
103817-804EA 7 USD
103817-804 103848-486 76721-616 76721-618 76721-620 77086-766 77086-768 77086-770
2-(1-Naphthalene)ethanol 97%
2-(1-Naphthalene)ethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₂O
MW: 172.23 g/mol
Boiling Pt: 311 °C (760 mm Hg)
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00004050
CAS Number: 773-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR