2-Methylthio-4-pyrimidinol 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Methylthio)pyrimidin-4(3H)-one, 2-(methylsulphanyl)-3,4-dihydropyrimidin-4-one, 2-Methylthio-4-pyrimidone, 4-Hydroxy-2-methylthiopyrimidine, S-Methyl-2-thiouracil

A129850-100G A129850-500G A129850-1G A129850-25G A129850-5G A129850-10G
103801-340EA 7 USD
103801-340 77285-272 77285-274 77313-736 77313-738 77313-740 77747-927
2-Methylthio-4-pyrimidinol 98%
2-Methylthio-4-pyrimidinol

Precaución: for Research Use Only

Formula: C₅H₆N₂OS
MW: 142.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047373
CAS Number: 5751-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR