2-(Methylsulfonyl)ethanamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Methylsulphonyl)ethanamine hydrochloride, 2-Aminoethyl Methyl sulphone Hydrochloride, 2-Aminoethyl methyll sulphone hydrochloride, 2-(Methylsulphonyl)ethanamine Hydrochloride

A176859-25G A176859-5G A176859-100G A176859-10G A176859-500G A176859-1G
103807-176EA 5 USD
103807-176 103845-504 77082-866 77082-868 77260-104 77260-106 77260-108 77301-470
2-(Methylsulfonyl)ethanamine hydrochloride 98%
2-(Methylsulphonyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₁₀ClNO₂S
MW: 159.6400 g/mol
Boiling Pt: 325.8 °C (760 mm Hg)
Flash Pt: 150.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03840162
CAS Number: 104458-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR