Methanesulfonylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Methylsulphonyl)acetic acid

A168286-10G A168286-1G A168286-25G A168286-5G
103806-266EA 6 USD
103806-266 103844-524 76799-294 77081-580 77081-582 77081-584
Methanesulfonylacetic acid 98%
Methanesulfonylacetic acid

Precaución: for Research Use Only

Formula: C₃H₆O₄S
MW: 138.14 g/mol
Boiling Pt: 423.6 °C (760 mm Hg)
Melting Pt: 118…120 °C
Flash Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00051843
CAS Number: 2516-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR