N-Methyl-N-phenylglycine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Methyl(phenyl)amino)acetic acid hydrochloride

A149625-10G A149625-250MG
103804-062EA 34 USD
103804-062 103842-184 76823-792 77885-236
N-Methyl-N-phenylglycine hydrochloride 97%
N-Methyl-N-phenylglycine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO₂
Boiling Pt: 311.6 °C (760 mm Hg)
Flash Pt: 142.3 °C
Storage Temperature: Ambient
MDL Number: MFCD13186269
CAS Number: 21911-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR