2-(Methoxymethoxy)-4-(trifluoromethoxy)benzaldehyde 95%

Supplier: Ambeed
Warning

A101997-250MG A101997-1G
103796-244EA 105 USD
103796-244 103821-940 76646-744 77308-822
2-(Methoxymethoxy)-4-(trifluoromethoxy)benzaldehyde 95%
2-(Methoxymethoxy)-4-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₄
MW: 250.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27943593
CAS Number: 1321963-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR