2-(Isopropylthio)aniline 95%

Supplier: Ambeed
Warning

A641613-1G A641613-25G A641613-5G A641613-100G A641613-10G
103840-342EA 65 USD
103840-342 76844-516 76844-518 76844-520 77018-290 77018-292
2-(Isopropylthio)aniline 95%
2-(Isopropylthio)aniline

Precaución: for Research Use Only

Formula: C₉H₁₃NS
Boiling Pt: 261.275 °C (760 mm Hg)
Flash Pt: 111.815 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08691961
CAS Number: 6397-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR