Iodomethylboronic acid, pinacol ester 95%

Supplier: Ambeed
Danger

Synonyms: 2-(Iodomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A362511-25G A362511-10G A362511-1G
103826-322EA 5 USD
103826-322 103837-338 76840-554 77167-862 77306-742
Iodomethylboronic acid, pinacol ester 95%
Iodomethylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₇H₁₄BIO₂
MW: 267.9 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12405515
CAS Number: 70557-99-2
UN: 1760
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR