2-(Hydroxymethyl)-1,3-propanediol 97%

Supplier: Ambeed
Warning

Synonyms: Tris(hydroxymethyl)methane, 2-(Hydroxymethyl)propane-1,3-diol

A169153-100G A169153-10G A169153-250MG A169153-1G
103806-356EA 16 USD
103806-356 103844-612 77247-926 77247-928 77247-930 77300-968
2-(Hydroxymethyl)-1,3-propanediol 97%
2-(Hydroxymethyl)-1,3-propanediol

Precaución: for Research Use Only

Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 294.9 °C (760 mm Hg)
Melting Pt: 63…68 °C
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00192217
CAS Number: 4704-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR