2-(Hydroxymethyl)isonicotinonitrile 95%

Supplier: Ambeed
Warning

A273954-100MG A273954-250MG A273954-1G
103813-170EA 110 USD
103813-170 103853-096 77289-474 77289-476 77305-154
2-(Hydroxymethyl)isonicotinonitrile 95%
2-(Hydroxymethyl)isonicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₆N₂O
Boiling Pt: 282.4 °C (760 mm Hg)
Flash Pt: 124.6 °C
Storage Temperature: Ambient
MDL Number: MFCD07774127
CAS Number: 51454-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR