2-(Hydroxymethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A169908-25G A169908-5G A169908-100G A169908-10G A169908-1G
103806-426EA 8 USD
103806-426 103844-692 77081-792 77081-794 77247-954 77247-956 77301-010
2-(Hydroxymethyl)benzoic acid 97%
2-(Hydroxymethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.147 g/mol
Boiling Pt: 333.9 °C (760 mm Hg)
Flash Pt: 169.9 °C
Storage Temperature: Ambient
MDL Number: MFCD02151998
CAS Number: 612-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR