2-(Hydroxymethyl)-3-methylphenol 95%

Supplier: Ambeed
Warning

A100374-100MG A100374-250MG
103795-404EA 113 USD
103795-404 103821-102 77008-268 77008-270
2-(Hydroxymethyl)-3-methylphenol 95%
2-(Hydroxymethyl)-3-methylphenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18397251
CAS Number: 29922-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR