SCH-58261 98%

Supplier: Ambeed
Warning

Synonyms: SCH 58261, 2-(furan-2-yl)-7-phenethyl-7H-pyrazolo[4,3-e][1,2,4]triazolo[1,5-c]pyrimidin-5-amine, SCH58261, SCH-58261

A144938-1MG A144938-10MG A144938-5MG
103791-486EA 65 USD
103791-486 103792-192 103795-028 103799-382 76719-478 77316-808 77316-810
SCH-58261 98%
SCH-58261

Precaución: for Research Use Only

Formula: C₁₈H₁₅N₇O
MW: 345.36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00933778
CAS Number: 160098-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR