2-(N-Ethylanilino)ethanol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Ethyl-N-(2-hydroxyethyl)aniline, Ethylphenylethanolamine, 2-(Ethyl(phenyl)amino)ethanol, N-Ethyl-N-2-ethanolaniline
A239861-100G
A239861-25G
103809-700EA
87
USD
103809-700
76874-390
77304-174
2-(N-Ethylanilino)ethanol 98%
2-(N-Ethylanilino)ethanol
Precaución: for Research Use Only
Formula:
C₁₀H₁₅NO Boiling Pt: 268 °C (760 mm Hg) Melting Pt: 36…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020575 CAS Number: 92-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...