2-(Diphenylphosphino)biphenyl 97%

Supplier: Ambeed
Warning

A176956-100G A176956-10G A176956-250MG A176956-1G A176956-25G A176956-5G
103807-188EA 6 USD
103807-188 103845-514 77029-284 77029-286 77029-288 77082-874 77082-876 77082-878
2-(Diphenylphosphino)biphenyl 97%
2-(Diphenylphosphino)biphenyl

Precaución: for Research Use Only

Formula: C₂₄H₁₉P
MW: 338.38 g/mol
Boiling Pt: 473.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11559063
CAS Number: 13885-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR