2-(Dimethylamino)pyrimidine-5-carbonitrile 95%

Supplier: Ambeed
Warning

A398061-100MG A398061-250MG A398061-1G
103827-012EA 63 USD
103827-012 76829-220 76829-222 76841-488
2-(Dimethylamino)pyrimidine-5-carbonitrile 95%
2-(Dimethylamino)pyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₈N₄
MW: 148.17 g/mol
Boiling Pt: 298.7±32.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD18651605
CAS Number: 55338-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR