Dimethylaminoacetyl chloride hydrochloride 95%

Supplier: Ambeed
Danger

Synonyms: 2-(Dimethylamino)acetyl chloride hydrochloride

A689408-1G A689408-5G
103840-572EA 37 USD
103840-572 76844-788 76844-790
Dimethylaminoacetyl chloride hydrochloride 95%
Dimethylaminoacetyl chloride hydrochloride

Precaución: for Research Use Only

Formula: (CH₃)₂NCH₂COCl·HCl
MW: 158.03 g/mol
Boiling Pt: 162 °C (760 mm Hg)
Flash Pt: 51.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02094025
CAS Number: 60853-81-8
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR