N,N-Dimethylglycine 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid

A314013-100G A314013-10G A314013-1G A314013-500G A314013-25G A314013-5G
103810-084EA 126 USD
103810-084 103835-936 77032-652 77032-654 77032-656 77032-658 77306-104 77306-106
N,N-Dimethylglycine 98%
N,N-Dimethylglycine

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Boiling Pt: 175.2 °C (760 mm Hg)
Melting Pt: 178…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00004283
CAS Number: 1118-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR