2-Difluoromethyl-1H-benzoimidazole 98%
Supplier: Ambeed
|
Warning
|
A168853-100G
A168853-10G
A168853-250MG
A168853-5G
103806-324EA
6
USD
103806-324
103844-580
76784-138
76784-140
76784-142
77081-646
2-Difluoromethyl-1H-benzoimidazole 98%
2-Difluoromethyl-1H-benzoimidazole
Precaución: for Research Use Only
Formula:
C₈H₆F₂N₂ Boiling Pt: 309.8 °C (760 mm Hg) Melting Pt: 150…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00463015 CAS Number: 705-09-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...