2-(Difluoromethoxy)pyridin-4-ol 98%

Supplier: Ambeed
Warning

A232472-100MG A232472-250MG A232472-1G
103810-836EA 85 USD
103810-836 103850-590 76826-510 76826-512 77303-940
2-(Difluoromethoxy)pyridin-4-ol 98%
2-(Difluoromethoxy)pyridin-4-ol

Precaución: for Research Use Only

Formula: C₆H₅F₂NO₂
MW: 161.106 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18261897
CAS Number: 205043-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR