2-(Difluoromethoxy)pyridin-4-amine 97%

Supplier: Ambeed
Warning

A292772-100MG A292772-250MG
103813-950EA 295 USD
103813-950 76988-680 76988-682
2-(Difluoromethoxy)pyridin-4-amine 97%
2-(Difluoromethoxy)pyridin-4-amine

Precaución: for Research Use Only

Formula: C₆H₆F₂N₂O
MW: 160.121 g/mol
Boiling Pt: 263.3±35.0 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18261290
CAS Number: 1211587-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR