2-(Difluoromethoxy)isonicotinic acid 95%

Supplier: Ambeed
Warning

A165096-1G A165096-5G
103805-946EA 100 USD
103805-946 103844-204 77300-766 77300-768
2-(Difluoromethoxy)isonicotinic acid 95%
2-(Difluoromethoxy)isonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₃
MW: 189.12 g/mol
Boiling Pt: 370.6±37.0 °C (760 mm Hg)
Flash Pt: 177.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18258474
CAS Number: 1211581-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR