2-(Dibenzylamino)propane-1,3-diol 97%

Supplier: Ambeed
Warning

A138024-10G A138024-250MG A138024-1G A138024-25G A138024-5G
103802-548EA 16 USD
103802-548 77010-216 77010-218 77298-950 77298-952 77298-954
2-(Dibenzylamino)propane-1,3-diol 97%
2-(Dibenzylamino)propane-1,3-diol

Precaución: for Research Use Only

Formula: C₁₇H₂₁NO₂
MW: 271.35 g/mol
Melting Pt: 78…81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11656822
CAS Number: 246232-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR