1-Cyclopentenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Cyclopent-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclopentene, 1-Cyclopenteneboronic acid Pinacol ester, 2-(1-Cyclopentenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A106873-100G A106873-10G A106873-1G A106873-5G A106873-25G
103797-092EA 7 USD
103797-092 103822-848 77257-476 77257-478 77309-708 77309-710 77570-412
1-Cyclopentenylboronic acid pinacol ester 98%
1-Cyclopentenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₉BO₂
MW: 194.0784 g/mol
Boiling Pt: 210 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08669391
CAS Number: 287944-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR