1-Cyclohexenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclohexeneboronic acid pinacol ester, 2-(Cyclohex-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Cyclohexene-1-boronic acid pinacol ester

A201110-250MG A201110-5G A201110-10G
103817-268EA 6 USD
103817-268 103847-904 76721-456 77085-980 77567-914
1-Cyclohexenylboronic acid pinacol ester 97%
1-Cyclohexenylboronic acid pinacol ester

Precaución: For research use only.

Formula: C₁₂H₂₁BO₂
MW: 208.1049 g/mol
Boiling Pt: 232 °C (760 mm Hg)
Flash Pt: 103.9 °C
Storage Temperature: Freezer
MDL Number: MFCD05663845
CAS Number: 141091-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR