cis-4-(Boc-amino)cyclohexaneacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: cis-[4-(Boc-amino)cyclohexyl]acetic acid, 2-(cis-4-((tert-Butoxycarbonyl)amino)cyclohexyl)acetic acid

A117103-250MG A117103-1G
103798-830EA 58 USD
103798-830 103824-706 76797-118 77297-512
cis-4-(Boc-amino)cyclohexaneacetic acid 97%
cis-4-(Boc-amino)cyclohexaneacetic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01862296
CAS Number: 327156-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR