2-(Chloromethyl)quinoline hydrochloride 97%

Supplier: Ambeed
Danger

A327418-100G A327418-10G A327418-25G A327418-1G
103825-466EA 6 USD
103825-466 103836-416 76773-020 76773-022 76839-382 77306-358
2-(Chloromethyl)quinoline hydrochloride 97%
2-(Chloromethyl)quinoline hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₉Cl₂N
MW: 214.10 g/mol
Boiling Pt: 288.8 °C (760 mm Hg)
Melting Pt: 183…187 °C
Flash Pt: 155.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00012734
CAS Number: 3747-74-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR