2-(Chloromethyl)-3-methylpyridine hydrochloride 95%
Supplier: Ambeed
|
Danger
|
A208343-10G
A208343-250MG
A208343-1G
A208343-5G
103817-870EA
76
USD
103817-870
103848-558
76814-502
76814-504
77086-870
77086-872
2-(Chloromethyl)-3-methylpyridine hydrochloride 95%
2-(Chloromethyl)-3-methylpyridine hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₈ClN·ClH MW: 178.059 g/mol Melting Pt: 159 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08669778 CAS Number: 4370-22-3 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...