2-(Chloromethyl)-3,5-dimethylpyridine hydrochloride 97%

Supplier: Ambeed
Danger

A326379-100MG A326379-250MG A326379-1G A326379-25G A326379-5G
103825-432EA 24 USD
103825-432 103836-380 76828-272 76828-274 76839-334 76839-336 76839-338
2-(Chloromethyl)-3,5-dimethylpyridine hydrochloride 97%
2-(Chloromethyl)-3,5-dimethylpyridine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀ClN·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD11112031
CAS Number: 73590-93-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR