2-(Carboxymethyl)-4-nitrobenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A165087-250MG
A165087-1G
A165087-5G
103805-944EA
43
USD
103805-944
103844-202
76690-010
77300-762
77300-764
2-(Carboxymethyl)-4-nitrobenzoic acid 95%
2-(Carboxymethyl)-4-nitrobenzoic acid
Precaución: for Research Use Only
Formula:
C₉H₇NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD07782180 CAS Number: 39585-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...