2-Picolyl bromide hydrobromide 95%

Supplier: Ambeed
Danger

Synonyms: 2-(Bromomethyl)pyridine hydrobromide

A249859-5G A249859-100G A249859-10G A249859-25G A249859-1G
103811-898EA 10 USD
103811-898 103851-736 77091-088 77238-254 77238-256 77238-258 77304-506
2-Picolyl bromide hydrobromide 95%
2-Picolyl bromide hydrobromide

Precaución: for Research Use Only

Formula: C₆H₇Br₂N
MW: 252.94 g/mol
Melting Pt: 149…152 °C
Flash Pt: 75.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01863544
CAS Number: 31106-82-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR