2-(Bromomethyl)-5-(trifluoromethyl)furan 95%

Supplier: Ambeed
Danger

A938822-1G A938822-5G A938822-10G
103841-426EA 35 USD
103841-426 76845-694 76845-696 77118-932
2-(Bromomethyl)-5-(trifluoromethyl)furan 95%
2-(Bromomethyl)-5-(trifluoromethyl)furan

Precaución: for Research Use Only

Formula: C₆H₄BrF₃O
MW: 228.9946 g/mol
Boiling Pt: 174.3 °C (760 mm Hg)
Flash Pt: 59.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03086219
CAS Number: 17515-77-4
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR