2-(Bromomethyl)-4-fluoro-1-nitrobenzene 95%

Supplier: Ambeed
Danger

A635721-100MG A635721-250MG
103840-312EA 650 USD
103840-312 76992-274 76992-276
2-(Bromomethyl)-4-fluoro-1-nitrobenzene 95%
2-(Bromomethyl)-4-fluoro-1-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09736331
CAS Number: 82420-35-7
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR