5-Fluoro-2-methoxybenzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Bromomethyl)-4-fluoro-1-methoxybenzene

A639325-1G A639325-25G A639325-5G A639325-100MG A639325-10G A639325-250MG
103840-330EA 26 USD
103840-330 76844-504 76844-506 76844-508 77117-558 77117-560 77117-562
5-Fluoro-2-methoxybenzyl bromide 98%
5-Fluoro-2-methoxybenzyl bromide

Precaución: for Research Use Only

Formula: C₈H₈BrFO
MW: 219.05 g/mol
Boiling Pt: 236.8 °C (760 mm Hg)
Melting Pt: 60…62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671769
CAS Number: 700381-18-6
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR