2-(Bromomethyl)-1-iodo-4-(trifluoromethyl)benzene 97%
Supplier: Ambeed
|
Warning
|
A182924-250MG
A182924-1G
103815-586EA
44
USD
103815-586
103846-148
76784-644
77083-726
2-(Bromomethyl)-1-iodo-4-(trifluoromethyl)benzene 97%
2-(Bromomethyl)-1-iodo-4-(trifluoromethyl)benzene
Precaución: for Research Use Only
Formula:
C₈H₅BrF₃I MW: 364.92 g/mol Boiling Pt: 268.3 °C (760 mm Hg) Flash Pt: 116 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11849937 CAS Number: 702641-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...