2-(Bromomethyl)-1,3-dimethylbenzene 97%

Supplier: Ambeed
Warning

A162489-250MG A162489-1G A162489-25G A162489-5G
103805-620EA 45 USD
103805-620 103843-870 77110-752 77319-912 77319-914 77319-916
2-(Bromomethyl)-1,3-dimethylbenzene 97%
2-(Bromomethyl)-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁Br
MW: 199.0876 g/mol
Boiling Pt: 237.8 °C (760 mm Hg)
Melting Pt: 33…37 °C
Flash Pt: 103.527 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03844742
CAS Number: 83902-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR